RuCl2[(R)-dm-segphos®][(R)-daipen]

Supplier: TCI America

Synonyms: Dichloro{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}[(2R)-(-)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) RuCl2[(R)-dm-segphos®][(R)-daipen]

R0129-1G R0129-200MG
TCR0129-1GEA 387.1 CAD
TCR0129-1G TCR0129-200MG
RuCl2[(R)-dm-segphos®][(R)-daipen]
RuCl2[(R)-dm-segphos®][(R)-daipen]

CAS Number: 944450-43-5
MDL Number: MFCD10567024
Molecular Formula: C65H70Cl2N2O6P2Ru
Molecular Weight: 1209.20
Form: Crystal
Color: Yellow

Formula: C₆₅H₇₀Cl₂N₂O₆P₂Ru
MW: 1209.20 g/mol
MDL Number: MFCD10567024
CAS Number: 944450-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e37eb78899d7d6a769a4b128c026e4d0a0028ed&vid=4e37eb78899d7d6a769a4b128c026e4d0a0028ed&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR