RuCl2[(S)-dm-segphos®][(S)-daipen]

Supplier: TCI America

Synonyms: Dichloro{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}[(2S)-(+)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) RuCl2[(S)-dm-segphos®][(S)-daipen]

R0128-1G R0128-200MG
TCR0128-1GEA 376.04 CAD
TCR0128-1G TCR0128-200MG
RuCl2[(S)-dm-segphos®][(S)-daipen]
RuCl2[(S)-dm-segphos®][(S)-daipen]

CAS Number: 944450-44-6
MDL Number: MFCD09753029
Molecular Formula: C65H70Cl2N2O6P2Ru
Molecular Weight: 1209.20
Form: Crystal
Color: Yellow

Formula: C₆₅H₇₀Cl₂N₂O₆P₂Ru
MW: 1209.20 g/mol
MDL Number: MFCD09753029
CAS Number: 944450-44-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb88d3dfba4dbb63a8f5a8b4317417fee73d420a&vid=cb88d3dfba4dbb63a8f5a8b4317417fee73d420a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR