RuCl[(R,R)-Tsdpen](mesitylene) ≥90% (by HPLC)

Supplier: TCI America

Synonyms: [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(mesitylene)ruthenium(II), [N-[(1R,2R)-2-(Amino-kN)-1,2- diphenylethyl]-4-methylbenzenesulfonamidato-kN]chloro [(1,2,3,4,5,6-n)-1,3,5-trimethylbenzene]-ruthenium, [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(mesitylene)ruthenium(II), [N-[(1R,2R)-2-(Amino-kN)-1,2- diphenylethyl]-4-methylbenzenesulphonamidato-kN]chloro [(1,2,3,4,5,6-n)-1,3,5-trimethylbenzene]-ruthenium, Chloro(mesitylene)[(R,R)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II), Chloro(mesitylene)[(R,R)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II)

R0127-1G R0127-200MG
TCR0127-1GEA 241.27 CAD
TCR0127-1G TCR0127-200MG
RuCl[(R,R)-Tsdpen](mesitylene) ≥90% (by HPLC)
RuCl[(R,R)-Tsdpen](mesitylene)

CAS Number: 174813-82-2
MDL Number: MFCD12545951
Molecular Formula: C30H33ClN2O2RuS
Molecular Weight: 622.19
Purity/Analysis Method: >90% (HPLC)
Form: Crystal
Color: Yellow


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(mesitylene)ruthenium(II), Chloro(mesitylene)[(R,R)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)

Formula: C₃₀H₃₃ClN₂O₂RuS
MW: 622.19 g/mol
MDL Number: MFCD12545951
CAS Number: 174813-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=786e23fcab6647577d947a91601d77d18b6b4cb2&vid=786e23fcab6647577d947a91601d77d18b6b4cb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR