RuCl[(S,S)-Tsdpen](mesitylene) ≥90%

Supplier: TCI America

Synonyms: [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(mesitylene)ruthenium(II), Chloro(mesitylene)[(S,S)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(mesitylene)ruthenium(II), [N-[(1S,2S)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulphonamidato-κN]chloro[(1,2,3,4,5,6-η)-1,3,5-trimethylbenzene]ruthenium, [N-[(1S,2S)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1,3,5-trimethylbenzene]ruthenium, Chloro(mesitylene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)

R0126-1G R0126-200MG
TCR0126-1GEA 256.28 CAD
TCR0126-1G TCR0126-200MG
RuCl[(S,S)-Tsdpen](mesitylene) ≥90%
RuCl[(S,S)-Tsdpen](mesitylene)

CAS Number: 174813-81-1
MDL Number: MFCD12545952
Molecular Formula: C30H33ClN2O2RuS
Molecular Weight: 622.19
Purity/Analysis Method: >90% (HPLC)
Form: Crystal
Color: Yellow


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(mesitylene)ruthenium(II), Chloro(mesitylene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)

Formula: C₃₀H₃₃ClN₂O₂RuS
MW: 622.19 g/mol
MDL Number: MFCD12545952
CAS Number: 174813-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93761b16b4b35d997698cd585caff007cdfd92fe&vid=93761b16b4b35d997698cd585caff007cdfd92fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR