RuCl[(R,R)-Tsdpen](p-cymene) ≥90% (by HPLC)

Supplier: TCI America

R0125-1G R0125-200MG
TCR0125-1GEA 229.73 CAD
TCR0125-1G TCR0125-200MG
RuCl[(R,R)-Tsdpen](p-cymene) ≥90% (by HPLC)
RuCl[(R,R)-Tsdpen](p-cymene)

CAS Number: 192139-92-7
MDL Number: MFCD10567385
Molecular Formula: C31H35ClN2O2RuS
Molecular Weight: 636.21
Purity/Analysis Method: >90% (HPLC)
Form: Crystal
Color: Yellow
Melting point (°C): 215
Specific rotation [a]20/D: -116 deg (C=0.1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)

Formula: C₃₁H₃₅ClN₂O₂RuS
Melting Pt: 215 °C
MDL Number: MFCD10567385
CAS Number: 192139-92-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f1dc31004440dc664187fb5515c120422364263&vid=1f1dc31004440dc664187fb5515c120422364263&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR