Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulfonyl)amido}(p-cymene)ruthenium(II) ≥90%

Supplier: TCI America

Synonyms: (S,S)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), RuCl[(S,S)-Tsdpen](p-cymene)

R0124-1G R0124-200MG
TCR0124-1GEA 305.7 CAD
TCR0124-1G TCR0124-200MG
Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulfonyl)amido}(p-cymene)ruthenium(II) ≥90%
Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulfonyl)amido}(p-cymene)ruthenium(II)

Form: Crystal
Color: Yellow
Specific rotation [a]20/D: 178 deg (C=0.5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)

Formula: C₃₁H₃₅ClN₂O₂RuS
MW: 636.21 g/mol
MDL Number: MFCD09953484
CAS Number: 192139-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83a865a06ef377080c334e52dcebbef8ec8ff684&vid=83a865a06ef377080c334e52dcebbef8ec8ff684&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR