RuCl[(R,R)-Fsdpen](p-cymene) ≥90% (by NMR)

Supplier: TCI America

R0123-1G R0123-200MG
TCR0123-1GEA 439.71 CAD
TCR0123-1G TCR0123-200MG
RuCl[(R,R)-Fsdpen](p-cymene) ≥90% (by NMR)
RuCl[(R,R)-Fsdpen](p-cymene)

CAS Number: 1026995-71-0
MDL Number: MFCD12545963
Molecular Formula: C30H28ClF5N2O2RuS
Molecular Weight: 712.14
Purity/Analysis Method: >90% (NMR)
Form: Crystal
Color: Yellow
Melting point (°C): 244
Flash Point (°C): 100
Specific rotation [a]20/D: -16 deg (C=0.1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: [(R,R)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)

Formula: C₃₀H₂₈ClF₅N₂O₂RuS
Melting Pt: 244 °C
Flash Pt: 100 °C
MDL Number: MFCD12545963
CAS Number: 1026995-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce16a7d4c37940c1d406dcb85f8e4ec195e2c169&vid=ce16a7d4c37940c1d406dcb85f8e4ec195e2c169&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR