RuCl[(S,S)-Fsdpen](p-cymene) ≥90%

Supplier: TCI America

Synonyms: Chloro(p-cymene)[(S,S)-N-(pentafluorobenzenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulphonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II), Chloro{[(1S,2S)-(-)-2-amino-1,2-diphenylethyl](pentafluorophenylsulfonyl)amido}(p-cymene)ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro{[(1S,2S)-(-)-2-amino-1,2-diphenylethyl](pentafluorophenylsulphonyl)amido}(p-cymene)ruthenium(II)

R0122-1G R0122-200MG
TCR0122-1GEA 439.71 CAD
TCR0122-1G TCR0122-200MG
RuCl[(S,S)-Fsdpen](p-cymene) ≥90%
RuCl[(S,S)-Fsdpen](p-cymene)

CAS Number: 1026995-72-1
MDL Number: MFCD12545954
Molecular Formula: C30H28ClF5N2O2RuS
Molecular Weight: 712.14
Purity/Analysis Method: >90% (NMR)
Form: Crystal
Color: Yellow
Melting point (°C): 244
Flash Point (°C): 100
Specific rotation [a]20/D: 48 deg (C=0.1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: [(S,S)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)

Formula: C₃₀H₂₈ClF₅N₂O₂RuS
MW: 712.14 g/mol
Melting Pt: 244 °C
Flash Pt: 100 °C
MDL Number: MFCD12545954
CAS Number: 1026995-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=018a54109d7b0eea56041e53aeed0eae4fbdf145&vid=018a54109d7b0eea56041e53aeed0eae4fbdf145&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR