Ranolazine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

R0112-1G R0112-5G
TCR0112-1GEA 289.3 CAD
TCR0112-1G TCR0112-5G
Ranolazine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Ranolazine dihydrochloride

CAS Number: 95635-56-6
MDL Number: MFCD03788770
Molecular Formula: C24H33N3O4
Molecular Weight: 500.46
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 227

Formula: C₂₄H₃₃N₃O₄
Melting Pt: 227 °C
MDL Number: MFCD03788770
CAS Number: 95635-56-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42d3464090952500d1223929d4335fd604c20355&vid=42d3464090952500d1223929d4335fd604c20355&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR