Rolipram ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

R0110-250MG R0110-50MG
TCR0110-250MGEA 1019.89 CAD
TCR0110-250MG TCR0110-50MG
Rolipram ≥98.0% (by HPLC, total nitrogen)
Rolipram

CAS Number: 61413-54-5
MDL Number: MFCD00270906
Molecular Formula: C16H21NO3
Molecular Weight: 275.35
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 132
Storage Temperature: >-20°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Neurotransmitter Agent Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-2-pyrrolidinone

Formula: C₁₆H₂₁NO₃
Melting Pt: 132 °C
MDL Number: MFCD00270906
CAS Number: 61413-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616c253617a86c2ead44de2b5aba318d969fa330&vid=616c253617a86c2ead44de2b5aba318d969fa330&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR