13-cis-Retinoic acid ≥97.0%
Supplier: TCI America
Synonyms:
Isotretinoin, (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
CAS Number: 4759-48-2
MDL Number: MFCD00079542
Molecular Formula: C20H28O2
Molecular Weight: 300.44
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 175
Lambda max.: 354 nm (EtOH)Storage Temperature: <0°C
Product Categories: Bioscience, Vitamins, Diterpenes
Other Product Names / Synonyms: (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, Isotretinoin
Formula:
C₂₀H₂₈O₂ Melting Pt: 175 °C |
MDL Number:
MFCD00079542 CAS Number: 4759-48-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...