Quinoxalin-5-ol ≥98.0%
Supplier: TCI America
Synonyms:
5-Hydroxyquinoxaline
Q0095-1G
Q0095-200MG
TCQ0095-1GEA
165.9
CAD
TCQ0095-1G
TCQ0095-200MG
Quinoxalin-5-ol ≥98.0%
Quinoxalin-5-ol
CAS Number: 17056-99-4
MDL Number: MFCD00972004
Molecular Formula: C8H6N2O
Molecular Weight: 146.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Pale Yellow
Boiling point (°C): 184
Melting point (°C): 101
Lambda max.: 250 nm (H2O)
Other Product Names / Synonyms: 5-Hydroxyquinoxaline
Formula:
C8H6N2O MW: 146.05 g/mol Boiling Pt: 184 °C Melting Pt: 101 °C |
MDL Number:
MFCD00972004 CAS Number: 17056-99-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...