3-Quinolinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Q0094-1G Q0094-5G
TCQ0094-1GEA 100.8 CAD
TCQ0094-1G TCQ0094-5G
3-Quinolinol ≥98.0% (by GC, titration analysis)
3-Quinolinol

CAS Number: 580-18-7
MDL Number: MFCD00169018
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 203


Other Product Names / Synonyms: 3-Hydroxyquinoline

Formula: C₉H₇NO
Melting Pt: 203 °C
MDL Number: MFCD00169018
CAS Number: 580-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6e0f0b5e6f04371bbdc9020e7aed71f675e0047&vid=c6e0f0b5e6f04371bbdc9020e7aed71f675e0047&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR