3-Quinolinecarboxylic acid ≥98.0%

Supplier: TCI America

Q0090-1G Q0090-5G
TCQ0090-1GEA 62.88 CAD
TCQ0090-1G TCQ0090-5G
3-Quinolinecarboxylic acid ≥98.0%
3-Quinolinecarboxylic acid

CAS Number: 6480-68-8
MDL Number: MFCD00006770
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 280

Formula: C₁₀H₇NO₂
MW: 173.17
Boiling Pt: 345
Melting Pt: 280 °C
MDL Number: MFCD00006770
CAS Number: 6480-68-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8262eb77dd2675c112ad8c0c41fa7ea476f590ad&vid=8262eb77dd2675c112ad8c0c41fa7ea476f590ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR