8-Quinolineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Q0086-1G Q0086-200MG
TCQ0086-1GEA 102.7 CAD
TCQ0086-1G TCQ0086-200MG
8-Quinolineboronic acid (contains varying amounts of Anhydride)
8-Quinolineboronic acid

CAS Number: 86-58-8
MDL Number: MFCD01114698
Molecular Formula: C9H8BNO2
Molecular Weight: 172.98
Form: Crystal
Color: White

Formula: C₉H₈BNO₂
MDL Number: MFCD01114698
CAS Number: 86-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=178c0400faec3de85c627f9fa8180455d6cffd72&vid=178c0400faec3de85c627f9fa8180455d6cffd72&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR