2-(1-Pyrazolyl)pyridine ≥98.0% (by GC)
Supplier: TCI America
P2239-1G
P2239-5G
TCP2239-1GEA
153.73
CAD
TCP2239-1G
TCP2239-5G
2-(1-Pyrazolyl)pyridine ≥98.0% (by GC)
2-(1-Pyrazolyl)pyridine
CAS Number: 25700-11-2
MDL Number: MFCD09033534
Molecular Formula: C8H7N3
Molecular Weight: 145.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 39
Formula:
C₈H₇N₃ Melting Pt: 39 °C |
MDL Number:
MFCD09033534 CAS Number: 25700-11-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...