Palladium 5% on alumina

Supplier: TCI America

P2238-25G P2238-5G
TCP2238-25GEA 938.84 CAD
TCP2238-25G TCP2238-5G
Palladium 5% on alumina
Palladium

CAS Number: 7440-05-3
Molecular Formula: Pd
Molecular Weight: 106.42
Form: Crystals
Color: Black


Product Categories: Synthetic Organic Chemistry, Metal Catalysts for Organic Synthesis, Suzuki-Miyaura Cross-coupling Reaction

Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bf65fcea6067db51024e04d9b9207fd001f2302&vid=6f3a44fc9ddd9a2caa64ca91fe91cfc6c584f537&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR