2-(1H-Pyrazol-3-yl)phenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(2-Hydroxyphenyl)pyrazole

P2218-1G P2218-5G
TCP2218-1GEA 104.28 CAD
TCP2218-1G TCP2218-5G
2-(1H-Pyrazol-3-yl)phenol ≥98.0% (by GC, titration analysis)
2-(1H-Pyrazol-3-yl)phenol

CAS Number: 34810-67-8
MDL Number: MFCD03093876
Molecular Formula: C9H8N2O
Molecular Weight: 160.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 97
Lambda max.: 294 nm (EtOH)


Other Product Names / Synonyms: 3-(2-Hydroxyphenyl)pyrazole

Formula: C₉H₈N₂O
MW: 160.18 g/mol
Melting Pt: 97 °C
MDL Number: MFCD03093876
CAS Number: 34810-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28eb6abe6bc581cae8a326a720a7d50b93401d6a&vid=1dc7b7f682b131c5002a6b53eaf84c6eefe81e14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR