2-Phenylethylamine hydroiodide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

P2213-1G P2213-5G
TCP2213-1GEA 165.9 CAD
TCP2213-1G TCP2213-5G
2-Phenylethylamine hydroiodide ≥98.0% (by HPLC, total nitrogen)
2-Phenylethylamine hydroiodide

CAS Number: 151059-43-7
Molecular Formula: C8H11N
Molecular Weight: 249.10
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 259


Product Categories: Materials Chemistry, Organic-Inorganic Perovskite Precursors
Other Product Names / Synonyms: Phenethylamine Hydroiodide

Formula: C₈H₁₁N
Melting Pt: 259 °C
CAS Number: 151059-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=095a442bfc44b23c25e5af1a142d0934e944eab5&vid=095a442bfc44b23c25e5af1a142d0934e944eab5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR