Propylamine hydroiodide ≥97.0%

Supplier: TCI America

P2212-1G P2212-5G
TCP2212-1GEA 79 CAD
TCP2212-1G TCP2212-5G
Propylamine hydroiodide ≥97.0%
Propylamine hydroiodide

CAS Number: 14488-45-0
Molecular Formula: C3H9N
Molecular Weight: 187.02
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal
Color: White

Formula: C₃H₉N
MW: 187.02 g/mol
CAS Number: 14488-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8abf4c0516148c9f48510d34ed98fc1fd16e55b&vid=e8abf4c0516148c9f48510d34ed98fc1fd16e55b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR