2-(1H-Pyrazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
P2211-1G
P2211-5G
TCP2211-1GEA
129.56
CAD
TCP2211-1G
TCP2211-5G
2-(1H-Pyrazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)
2-(1H-Pyrazol-3-yl)pyridine
CAS Number: 75415-03-1
MDL Number: MFCD00115151
Molecular Formula: C8H7N3
Molecular Weight: 145.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 124
Lambda max.: 303 nm (CH3CN aq.)
Other Product Names / Synonyms: 3-(2-Pyridyl)pyrazol
Formula:
C₈H₇N₃ Melting Pt: 124 °C |
MDL Number:
MFCD00115151 CAS Number: 75415-03-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...