2-(2-Propynyloxy)naphthalene ≥98.0%

Supplier: TCI America

Synonyms: 2-(Prop-2-yn-1-yloxy)naphthalene, 2-(Propargyloxy)naphthalene

P2190-1G P2190-200MG
TCP2190-1GEA 654.59 CAD
TCP2190-1G TCP2190-200MG
2-(2-Propynyloxy)naphthalene ≥98.0%
2-(2-Propynyloxy)naphthalene

CAS Number: 20009-28-3
Molecular Formula: C13H10O
Molecular Weight: 182.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 83
Melting point (°C): 60


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 2-(Propargyloxy)naphthalene

Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 83 °C
Melting Pt: 60 °C
MDL Number: MFCD04275627
CAS Number: 20009-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d83fc6fceb6cd33bb7bc4e314e7adb8bba552b90&vid=d83fc6fceb6cd33bb7bc4e314e7adb8bba552b90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR