Pentafluorophenyl trifluoromethanesulfonate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Trifluoromethanesulphonic acid Pentafluorophenyl Ester, Perfluorophenyl trifluoromethanesulphonate, Trifluoromethanesulfonic acid Pentafluorophenyl Ester, Pentafluorophenyl Triflate, Perfluorophenyl trifluoromethanesulfonate

P2188-1G P2188-200MG
TCP2188-1GEA 215.43 CAD
TCP2188-1G TCP2188-200MG
Pentafluorophenyl trifluoromethanesulfonate ≥97.0% (by GC)
Pentafluorophenyl trifluoromethanesulfonate

CAS Number: 60129-85-3
MDL Number: MFCD07784319
Molecular Formula: C7F8O3S
Molecular Weight: 316.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 40
Specific Gravity (20/20): 1.75


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Triflation, Protection & Derivatization Reagents (for Synthesis), Fluorinated Building Blocks
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid Pentafluorophenyl Ester, Pentafluorophenyl Triflate

Formula: C₇F₈O₃S
MW: 316.12 g/mol
Boiling Pt: 40 °C
Density: 1.75 (20 °C)
MDL Number: MFCD07784319
CAS Number: 60129-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14e89e3c47e41b061e2f8691632f27d1834c2221&vid=14e89e3c47e41b061e2f8691632f27d1834c2221&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR