Pentafluorophenyl acrylate ≥98.0%

Supplier: TCI America

Synonyms: Acrylic Acid Pentafluorophenyl Ester

P2179-1G P2179-5G
TCP2179-1GEA 158 CAD
TCP2179-1G TCP2179-5G
Pentafluorophenyl acrylate ≥98.0%
Pentafluorophenyl acrylate

CAS Number: 71195-85-2
MDL Number: MFCD00042330
Molecular Formula: C9H3F5O2
Molecular Weight: 238.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 197
Flash Point (°C): 68
Specific Gravity (20/20): 1.46
Lambda max.: 256 nm (Hexane)Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Pentafluorophenyl Ester

Formula: C₉H₃F₅O₂
MW: 238.11 g/mol
Boiling Pt: 197 °C
Density: 1.46 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00042330
CAS Number: 71195-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1baee82d513c9d91f0648d97ae52fe97c3c547d&vid=f1baee82d513c9d91f0648d97ae52fe97c3c547d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR