(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) ≥95.0%
Supplier: TCI America
P2172-1G
P2172-5G
TCP2172-1GEA
628.84
CAD
TCP2172-1G
TCP2172-5G
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) ≥95.0%
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I)
CAS Number: 1334890-93-5
Molecular Formula: C31H23CuF3N2P
Molecular Weight: 575.05
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: Pale Yellow
Formula:
C₃₁H₂₃CuF₃N₂P MW: 575.05 g/mol |
MDL Number:
MFCD28386111 CAS Number: 1334890-93-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...