9-Phenylcarbazole-2-boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(9-Phenyl-9H-carbazol-2-yl)boronic acid
P2169-1G
P2169-200MG
TCP2169-1GEA
100.8
CAD
TCP2169-1G
TCP2169-200MG
9-Phenylcarbazole-2-boronic acid (contains varying amounts of Anhydride)
9-Phenylcarbazole-2-boronic acid
CAS Number: 1001911-63-2
MDL Number: MFCD22207050
Molecular Formula: C18H14BNO2
Molecular Weight: 287.13
Form: Crystal
Color: White
Formula:
C₁₈H₁₄BNO₂ MW: 287.13 g/mol |
MDL Number:
MFCD22207050 CAS Number: 1001911-63-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...