Nicotinamidine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

P2167-1G P2167-5G
TCP2167-1GEA 75.84 CAD
TCP2167-1G TCP2167-5G
Nicotinamidine hydrochloride ≥98.0% (by HPLC)
Nicotinamidine hydrochloride

CAS Number: 7356-60-7
MDL Number: MFCD00054326
Molecular Formula: C6H7N3
Molecular Weight: 157.60
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 189


Other Product Names / Synonyms: 3-Carbamimidoylpyridine Monohydrochloride

Formula: C₆H₈ClN₃
Melting Pt: 189 °C
MDL Number: MFCD00054326
CAS Number: 7356-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b463c2efe145b712bb1775e16e5e0b8a8b44720f&vid=bea789b0c6dd7b1b5289b9a93f1ef57e4df3605c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR