Propylene glycol monostearate ≥83.0% (by GC)

Supplier: TCI America

Synonyms: Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester

P2163-25G P2163-500G
TCP2163-25GEA 54.51 CAD
TCP2163-25G TCP2163-500G
Propylene glycol monostearate ≥83.0% (by GC)
Propylene glycol monostearate

CAS Number: 1323-39-3
Molecular Formula: C21H42O3
Molecular Weight: 342.56
Purity/Analysis Method: >83.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 44


Other Product Names / Synonyms: Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester

Formula: C₂₁H₄₂O₃
MW: 342.56 g/mol
Melting Pt: 44 °C
MDL Number: MFCD00021902
CAS Number: 1323-39-3

Order Now

Specification Test Results

Purity/Analysis Method 83.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR