4-(trans-4-Propylcyclohexyl)phenylboronic acid 97-108% (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (4-(trans-4-propylcyclohexyl)phenyl)boronic acid

P2162-1G P2162-5G
TCP2162-1GEA 47.4 CAD
TCP2162-1G TCP2162-5G
4-(trans-4-Propylcyclohexyl)phenylboronic acid 97-108% (contains varying amounts of Anhydride)
4-(trans-4-Propylcyclohexyl)phenylboronic acid

CAS Number: 146862-02-4
MDL Number: MFCD06658197
Molecular Formula: C15H23BO2
Molecular Weight: 246.16
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-(trans-4-Propylcyclohexyl)benzeneboronic Acid

Formula: C₁₅H₂₃BO₂
MW: 246.1529 g/mol
MDL Number: MFCD06658197
CAS Number: 146862-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b384ae047ffeafa897669aabd413aea4277dab6d&vid=b384ae047ffeafa897669aabd413aea4277dab6d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR