4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

P2158-1G P2158-5G
TCP2158-1GEA 139.04 CAD
TCP2158-1G TCP2158-5G
4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid

CAS Number: 952514-79-3
MDL Number: MFCD11045057
Molecular Formula: C19H15BN2O2
Molecular Weight: 314.15
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-(1-Phenyl-1H-benzimidazol-2-yl)benzeneboronic Acid

Formula: C₁₉H₁₅BN₂O₂
MDL Number: MFCD11045057
CAS Number: 952514-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41bb3539c18b6eb6bd5a2eadf57cf98bad440257&vid=41bb3539c18b6eb6bd5a2eadf57cf98bad440257&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR