Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate ≥97.0%

Supplier: TCI America

Synonyms: Potassium (((tert-butoxycarbonyl)amino)methyl)trifluoroborate, Potassium (N-Boc-aminomethyl)trifluoroborate

P2155-1G P2155-5G
TCP2155-1GEA 209.98 CAD
TCP2155-1G TCP2155-5G
Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate ≥97.0%
Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate

CAS Number: 1314538-55-0
MDL Number: MFCD19686142
Molecular Formula: C6H12BF3KNO2
Molecular Weight: 237.07
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Color: White
Melting point (°C): 181


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Potassium (N-Boc-aminomethyl)trifluoroborate

Formula: C₆H₁₂BF₃KNO₂
MW: 237.07 g/mol
Melting Pt: 181 °C
MDL Number: MFCD19686142
CAS Number: 1314538-55-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00de58c01555f419483eeb8565358b3cec168ded&vid=00de58c01555f419483eeb8565358b3cec168ded&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR