(R)-1-Phenyl-1,2-ethanediyl bis[4-(trans-4-pentylcyclohexyl)benzoate] ≥98.0%
Supplier: TCI America
Synonyms:
(R)-1-Phenylethane-1,2-diyl bis(4-((trans-4-pentylcyclohexyl)benzoate)
P2150-1G
P2150-200MG
TCP2150-1GEA
532.3
CAD
TCP2150-1G
TCP2150-200MG
(R)-1-Phenyl-1,2-ethanediyl bis[4-(trans-4-pentylcyclohexyl)benzoate] ≥98.0%
(R)-1-Phenyl-1,2-ethanediyl bis[4-(trans-4-pentylcyclohexyl)benzoate]
CAS Number: 154102-21-3
MDL Number: MFCD26961137
Molecular Formula: C44H58O4
Molecular Weight: 650.94
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 132
Formula:
C₄₄H₅₈O₄ Melting Pt: 132 °C |
MDL Number:
MFCD26961137 CAS Number: 154102-21-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...