4-[(1-Pyrrolidinylsulfonyl)methyl]aniline ≥97.0%

Supplier: TCI America

Synonyms: 4-((Pyrrolidin-1-ylsulphonyl)methyl)aniline, 1-[(4-Aminobenzyl)sulfonyl]pyrrolidine, 1-[(4-Aminobenzyl)sulphonyl]pyrrolidine, 4-((Pyrrolidin-1-ylsulfonyl)methyl)aniline

P2148-1G P2148-5G
TCP2148-1GEA 60.04 CAD
TCP2148-1G TCP2148-5G
4-[(1-Pyrrolidinylsulfonyl)methyl]aniline ≥97.0%
4-[(1-Pyrrolidinylsulfonyl)methyl]aniline

CAS Number: 334981-10-1
MDL Number: MFCD06795500
Molecular Formula: C11H16N2O2S
Molecular Weight: 240.32
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 178


Other Product Names / Synonyms: 1-[(4-Aminobenzyl)sulfonyl]pyrrolidine

Formula: C₁₁H₁₆N₂O₂S
MW: 240.32 g/mol
Melting Pt: 178 °C
MDL Number: MFCD06795500
CAS Number: 334981-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a90b0e472d7aab5e4356f25d5fcc5551490903c9&vid=a90b0e472d7aab5e4356f25d5fcc5551490903c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR