Pidotimod ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P2147-25G P2147-5G
TCP2147-25GEA 1414.29 CAD
TCP2147-25G TCP2147-5G
Pidotimod ≥98.0% (by HPLC, titration analysis)
Pidotimod

CAS Number: 121808-62-6
MDL Number: MFCD00867583
Molecular Formula: C9H12N2O4S
Molecular Weight: 244.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 187
Specific rotation [a]20/D: -136 deg (C=2, 5mol/L HCl)


Product Categories: Bioscience, Immunosuppressant Ingredients
Other Product Names / Synonyms: (R)-3-[[(S)-5-Oxo-2-pyrrolidinyl]carbonyl]-1,3-thiazolidine-4-carboxylic Acid

Formula: C₉H₁₂N₂O₄S
Melting Pt: 187 °C
MDL Number: MFCD00867583
CAS Number: 121808-62-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b1f08701c12343df828d7d22c481aa6ac6450a8&vid=4b1f08701c12343df828d7d22c481aa6ac6450a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR