2-Amino-6-phenylpyrimidin-4(3H)-one ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
2-Amino-6-phenyl-4(1H)-pyrimidinone, 6-Phenylisocytosine
P2123-1G
P2123-5G
TCP2123-1GEA
173.48
CAD
TCP2123-1G
TCP2123-5G
2-Amino-6-phenylpyrimidin-4(3H)-one ≥98.0% (by HPLC)
2-Amino-6-phenylpyrimidin-4(3H)-one
CAS Number: 56741-94-7
MDL Number: MFCD00466412
Molecular Formula: C10H9N3O
Molecular Weight: 187.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 295
Other Product Names / Synonyms: 2-Amino-6-phenyl-4(1H)-pyrimidinone
Formula:
C₁₀H₉N₃O MW: 187.2000 g/mol Melting Pt: 295 °C |
MDL Number:
MFCD00466412 CAS Number: 56741-94-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...