(2-Phenylethyl)phosphonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Phenethylphosphonic Acid

P2122-1G P2122-200MG
TCP2122-1GEA 1224.5 CAD
TCP2122-1G TCP2122-200MG
(2-Phenylethyl)phosphonic acid ≥98.0% (by HPLC, titration analysis)
(2-Phenylethyl)phosphonic acid

CAS Number: 4672-30-4
MDL Number: MFCD00485102
Molecular Formula: C8H11O3P
Molecular Weight: 186.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 138


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: Phenethylphosphonic Acid

Formula: C₈H₁₁O₃P
MW: 186.15 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00485102
CAS Number: 4672-30-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a498b76a37caa23ceaf1b66b725e58814bab0ad7&vid=a498b76a37caa23ceaf1b66b725e58814bab0ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR