Pranlukast ≥98.0% (by HPLC)

Supplier: TCI America

P2115-100MG P2115-25MG
TCP2115-100MGEA 173.58 CAD
TCP2115-100MG TCP2115-25MG
Pranlukast ≥98.0% (by HPLC)
Pranlukast

CAS Number: 103177-37-3
MDL Number: MFCD00864631
Molecular Formula: C27H23N5O4
Molecular Weight: 481.51
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 233
Storage Temperature: 0-10°C

Formula: C₂₇H₂₃N₅O₄
Melting Pt: 233 °C
MDL Number: MFCD00864631
CAS Number: 103177-37-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0680804e9c13baac60b45478f2894f148c1a3608&vid=0680804e9c13baac60b45478f2894f148c1a3608&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR