(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
(S)-1-(4-Fluorophenyl)-5-(2-oxo-4-phenyloxazolidin-3-yl)pentane-1,5-dione
P2111-25G
P2111-5G
TCP2111-25GEA
442.4
CAD
TCP2111-25G
TCP2111-5G
(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone
CAS Number: 189028-93-1
MDL Number: MFCD08061377
Molecular Formula: C20H18FNO4
Molecular Weight: 355.37
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 92
Formula:
C₂₀H₁₈FNO₄ MW: 355.37 g/mol Melting Pt: 92 °C |
MDL Number:
MFCD08061377 CAS Number: 189028-93-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...