3-Perylenecarboxaldehyde ≥95.0%

Supplier: TCI America

Synonyms: 3-Formylperylene

P2110-1G P2110-5G
TCP2110-1GEA 267.02 CAD
TCP2110-1G TCP2110-5G
3-Perylenecarboxaldehyde ≥95.0%
3-Perylenecarboxaldehyde

CAS Number: 35438-63-2
MDL Number: MFCD00127748
Molecular Formula: C21H12O
Molecular Weight: 280.33
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 235
Lambda max.: 476 nm (CHCl3)


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 3-Formylperylene

Formula: C₂₁H₁₂O
MW: 280.33 g/mol
Melting Pt: 235 °C
MDL Number: MFCD00127748
CAS Number: 35438-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43c9373a52b03fca3a53c5d29e59bce1be085501&vid=43c9373a52b03fca3a53c5d29e59bce1be085501&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR