N-(3-Pyridyl)pivalamide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,2-Dimethyl-N-pyridin-3-yl-propionamide, 2,2-Dimehtyl-N-pyridin-3-yl-propionamide
P2109-25G
P2109-5G
TCP2109-25GEA
247.9
CAD
TCP2109-25G
TCP2109-5G
N-(3-Pyridyl)pivalamide ≥98.0% (by GC, titration analysis)
N-(3-Pyridyl)pivalamide
CAS Number: 70298-88-3
MDL Number: MFCD00996243
Molecular Formula: C10H14N2O
Molecular Weight: 178.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 130
Other Product Names / Synonyms: 2,2-Dimethyl-N-(3-pyridyl)propionamide
Formula:
C₁₀H₁₄N₂O MW: 178.23 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD00996243 CAS Number: 70298-88-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...