4-(1-Pyrrolidinylcarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-(1-Pyrrolidinylcarbonyl)benzeneboronic acid, 4-(Pyrrolidine-1-carbonyl)phenylboronic acid, (4-(Pyrrolidine-1-carbonyl)phenyl)boronic acid

P2105-1G P2105-5G
TCP2105-1GEA 110.6 CAD
TCP2105-1G TCP2105-5G
4-(1-Pyrrolidinylcarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Pyrrolidinylcarbonyl)phenylboronic acid

CAS Number: 389621-81-2
MDL Number: MFCD03411950
Molecular Formula: C11H14BNO3
Molecular Weight: 219.05
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-[(1-Pyrrolidinyl)carbonyl]benzeneboronic Acid

Formula: C₁₁H₁₄BNO₃
MDL Number: MFCD03411950
CAS Number: 389621-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2dbefcd5c26e5c6e046d42ea627b9a76542ca31&vid=f2dbefcd5c26e5c6e046d42ea627b9a76542ca31&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR