Pyromellitic dianhydride ≥99.0% (by HPLC, titration analysis), purified by sublimation

Supplier: TCI America

P2103-25G P2103-5G
TCP2103-25GEA 684.14 CAD
TCP2103-25G TCP2103-5G
Pyromellitic dianhydride ≥99.0% (by HPLC, titration analysis), purified by sublimation
Pyromellitic dianhydride

CAS Number: 89-32-7
MDL Number: MFCD00005005
Molecular Formula: C10H2O6
Molecular Weight: 218.12
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Color: White
Boiling point (°C): 400
Melting point (°C): 286
Flash Point (°C): 167


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Highly Purified Reagents
Other Product Names / Synonyms: Benzene-1,2,4,5-tetracarboxylic Dianhydride

Formula: C₁₀H₂O₆
Boiling Pt: 400 °C
Melting Pt: 286 °C
Flash Pt: 167 °C
MDL Number: MFCD00005005
CAS Number: 89-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebe382e4ee1069a9e0120f7c10d403e4cb075516&vid=d376b376a2f6482671f4785d92cbc22e10739b51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR