2-(1H-1,2,4-Triazol-3-yl)pyridine ≥98.0%

Supplier: TCI America

Synonyms: 3-(2-Pyridyl)-1,2,4-triazole

P2100-1G P2100-200MG
TCP2100-1GEA 213.3 CAD
TCP2100-1G TCP2100-200MG
2-(1H-1,2,4-Triazol-3-yl)pyridine ≥98.0%
2-(1H-1,2,4-Triazol-3-yl)pyridine

CAS Number: 23195-62-2
MDL Number: MFCD13188589
Molecular Formula: C7H6N4
Molecular Weight: 146.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 162
Lambda max.: 280 nm


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 3-(2-Pyridyl)-1,2,4-triazole

Formula: C₇H₆N₄
Melting Pt: 162 °C
MDL Number: MFCD13188589
CAS Number: 23195-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8caa471414525831f602add90fb0733dd55ebd7&vid=c8caa471414525831f602add90fb0733dd55ebd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR