2-(2-Pyridyl)benzoxazole ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Pyridin-2-yl)benzo[d]oxazole

P2098-1G P2098-5G
TCP2098-1GEA 67.94 CAD
TCP2098-1G TCP2098-5G
2-(2-Pyridyl)benzoxazole ≥97.0% (by GC, titration analysis)
2-(2-Pyridyl)benzoxazole

CAS Number: 32959-62-9
MDL Number: MFCD00569582
Molecular Formula: C12H8N2O
Molecular Weight: 196.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 150
Melting point (°C): 103
Lambda max.: 302 nm (EtOH aq.)

Formula: C₁₂H₈N₂O
MW: 196.21 g/mol
Boiling Pt: 150 °C
Melting Pt: 103 °C
MDL Number: MFCD00569582
CAS Number: 32959-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38042abb1ef5f8eeece70354c70335d96e4121a5&vid=38042abb1ef5f8eeece70354c70335d96e4121a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR