Phenylmaleic anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
P2096-1G
P2096-5G
TCP2096-1GEA
143.78
CAD
TCP2096-1G
TCP2096-5G
Phenylmaleic anhydride ≥98.0% (by GC, titration analysis)
Phenylmaleic anhydride
CAS Number: 36122-35-7
MDL Number: MFCD00015472
Molecular Formula: C10H6O3
Molecular Weight: 174.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 122
Other Product Names / Synonyms: 3-Phenyl-2,5-furandione
Formula:
C₁₀H₆O₃ Melting Pt: 122 °C |
MDL Number:
MFCD00015472 CAS Number: 36122-35-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...