Potassium-p-tolyl sulfate ≥98.0%
Supplier: TCI America
Synonyms:
sulfuric acid p-Cresyl Ester Potassium Salt, p-Tolyl sulfate Potassium Salt, sulphuric acid p-Tolyl Ester Potassium Salt, sulphuric acid p-Cresyl Ester Potassium Salt, Potassium p-Cresyl sulfate, p-Cresyl sulphate potassium, sulfuric acid p-Tolyl Ester Potassium Salt, p-Cresyl sulfate potassium, p-Tolyl sulphate Potassium Salt, Potassium p-Cresyl Sulphate
CAS Number: 91978-69-7
Molecular Formula: C7H8O4S
Molecular Weight: 226.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Storage Temperature: <0°C
Other Product Names / Synonyms: Potassium p-Cresyl Sulfate, Sulfuric Acid p-Tolyl Ester Potassium Salt, Sulfuric Acid p-Cresyl Ester Potassium Salt
Formula:
C₇H₈O₄S MW: 226.29 g/mol |
MDL Number:
MFCD28100835 CAS Number: 91978-69-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...