Phenethyl caffeate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3,4-Dihydroxycinnamic Acid Phenethyl Ester, Caffeic Acid Phenethyl Ester, Phenethyl 3-(3,4-dihydroxyphenyl)acrylate, CAPE, Phenethyl 3,4-Dihydroxycinnamate

P2088-250MG P2088-25MG
TCP2088-250MGEA 189.71 CAD
TCP2088-250MG TCP2088-25MG
Phenethyl caffeate ≥98.0% (by HPLC)
Phenethyl caffeate

CAS Number: 104594-70-9
MDL Number: MFCD00866470
Molecular Formula: C17H16O4
Molecular Weight: 284.31
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 129
Lambda max.: 323 nmStorage Temperature: <0°C


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Regenerative Medicine Research Reagents, Antioxidants, Phenylpropanoids, Phenylpro
Other Product Names / Synonyms: Caffeic Acid Phenethyl Ester, Phenethyl 3,4-Dihydroxycinnamate, 3,4-Dihydroxycinnamic Acid Phenethyl Ester, CAPE

Formula: C₁₇H₁₆O₄
MW: 284.31 g/mol
Melting Pt: 129 °C
MDL Number: MFCD00866470
CAS Number: 104594-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9802c2d652adec592de77c3d4aa23a7a9dc0aba&vid=d9802c2d652adec592de77c3d4aa23a7a9dc0aba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR