Pyridine iodine monochloride ≥98.0%

Supplier: TCI America

Synonyms: Pyridine Iodochloride

P2086-1G P2086-5G
TCP2086-1GEA 237 CAD
TCP2086-1G TCP2086-5G
Pyridine iodine monochloride ≥98.0%
Pyridine iodine monochloride

CAS Number: 6443-90-9
MDL Number: MFCD06410987
Molecular Formula: C5H5N
Molecular Weight: 241.46
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 135
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: Pyridine Iodochloride

Formula: C₅H₅N
MW: 241.46 g/mol
Melting Pt: 135 °C
MDL Number: MFCD06410987
CAS Number: 6443-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=621ae8eba913ce323c1d1608f7a4df71096c289d&vid=621ae8eba913ce323c1d1608f7a4df71096c289d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR