Pentafluoro-L-phenylalanine ≥95.0%

Supplier: TCI America

Synonyms: (S)-2-Amino-3-(perfluorophenyl)propanoic acid

P2085-1G P2085-200MG
TCP2085-1GEA 494.54 CAD
TCP2085-1G TCP2085-200MG
Pentafluoro-L-phenylalanine ≥95.0%
Pentafluoro-L-phenylalanine

CAS Number: 34702-59-5
MDL Number: MFCD01860881
Molecular Formula: C9H6F5NO2
Molecular Weight: 255.14
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 258
Specific rotation [a]20/D: 19 deg (C=0.2, H2O)

Formula: C₉H₆F₅NO₂
Melting Pt: 258 °C
MDL Number: MFCD01860881
CAS Number: 34702-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3f158e0f469a5e7801f6e4206fdfdbf8edd3fa6&vid=b3f158e0f469a5e7801f6e4206fdfdbf8edd3fa6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR