Pentaerythritol tetraacrylate ≥80.0% (by GC) stabilized

Supplier: TCI America

P2084-25G P2084-500G
TCP2084-25GEA 59.57 CAD
TCP2084-25G TCP2084-500G
Pentaerythritol tetraacrylate ≥80.0% (by GC) stabilized
Pentaerythritol tetraacrylate

(stabilized with MEHQ)
CAS Number: 4986-89-4
MDL Number: MFCD00015334
Molecular Formula: C17H20O8
Molecular Weight: 352.34
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 113
Specific Gravity (20/20): 1.18

Formula: C₁₇H₂₀O₈
MW: 352.34 g/mol
Density: 1.18 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00015334
CAS Number: 4986-89-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50c42887097a03e99d7241ab8af4ebd2c3adfc37&vid=50c42887097a03e99d7241ab8af4ebd2c3adfc37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR